Assay Details
| Gene Symbol: Dusp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028099.4 | NP_082375.4 | 7 - 8 | 746 | 68 | |
| GenBank mRNA | AK008492.1 | - | 2 - 3 | 130 | 68 | |
| AK081629.1 | - | 7 - 8 | 723 | 68 | ||
| AK088941.1 | - | 7 - 8 | 743 | 68 | ||
| BC028640.1 | - | 3 - 4 | 173 | 68 | ||
| BC067028.1 | - | 7 - 8 | 700 | 30364073 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dusp11
Genomic Map