Assay Details
| Gene Symbol: Rpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042556.1 | NP_001036021.1 | 8 - 9 | 1015 | 79 | |
| NM_023323.3 | NP_075812.3 | 8 - 9 | 665 | 79 | ||
| GenBank mRNA | AB041810.1 | - | 8 - 9 | 1045 | 79 | |
| AK010776.1 | - | 8 - 9 | 659 | 79 | ||
| AK011644.1 | - | 8 - 9 | 643 | 79 | ||
| AK012188.1 | - | 8 - 9 | 665 | 79 | ||
| AK012641.1 | - | 8 - 9 | 644 | 79 | ||
| AK013395.1 | - | 8 - 9 | 640 | 79 | ||
| AK017678.1 | - | 8 - 9 | 636 | 79 | ||
| AK020030.1 | - | 8 - 9 | 626 | 79 | ||
| AK051135.1 | - | 8 - 9 | 1016 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpf2
Genomic Map