Assay Details
| Gene Symbol: Gm36595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK146861.1 | - | 319 | 138 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
gap junction protein, alpha 1 |
Gja1 |
AU042049, AW546267, Cnx43, Cx43, Cx43alpha1, Gja-1, Npm1, connexin43 |
-48.1kb | Sense | Chr. 10 - 56377201 - 56390423 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930467K11 gene | 4930467K11Rik | 1.0mb | Anti-sense | Chr. 10 - 57478383 - 57486353 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm36595
Genomic Map