Assay Details
| Gene Symbol: Lin28b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031772.2 | NP_001026942.1 | 5 | 4652 | 106 | |
| GenBank mRNA | AK050968.1 | - | 1 | 637 | 106 | |
| BC085193.1 | - | 3 | 4396 | 106 | ||
| BC089037.1 | - | 5 | 4643 | 30690977 | 106 | |
| DQ127225.1 | - | 5 | 4653 | 106 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lin28b
Genomic Map