Assay Details
| Gene Symbol: Mcm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027830.2 | NP_082106.2 | 5 - 6 | 1162 | 98 | |
| GenBank mRNA | AK018494.1 | - | 4 - 5 | 1002 | 98 | |
| AK046636.1 | - | 4 - 5 | 946 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm9
Genomic Map