Assay Details
| Gene Symbol: Lin28b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031772.2 | NP_001026942.1 | 3 - 4 | 516 | 74 | |
| GenBank mRNA | AK133928.1 | - | 3 - 4 | 535 | 74 | |
| BC089037.1 | - | 3 - 4 | 507 | 30690977 | 74 | |
| DQ127225.1 | - | 3 - 4 | 516 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lin28b
Genomic Map