Assay Details
| Gene Symbol: Slfnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177570.3 | NP_808238.1 | 1 - 2 | 254 | 64 | |
| GenBank mRNA | AK077106.1 | - | 1 - 2 | 255 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077106.1, NM_177570.3 transcripts.
Set Membership: |
|

Slfnl1
Genomic Map