Assay Details
| Gene Symbol: Slfnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177570.3 | NP_808238.1 | 4 - 5 | 1500 | 92 | |
| GenBank mRNA | AK077106.1 | - | 4 - 5 | 1501 | 92 | |
| BC117545.1 | - | 3 - 4 | 1248 | 40105640 | 92 | |
| BC118052.1 | - | 3 - 4 | 1251 | 40105639 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149687 | BC117545.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149687 | BC118052.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149687 | AK077106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149687 | NM_177570.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149687 | BC117545.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149687 | BC118052.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149687 | AK077106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149687 | NM_177570.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477817 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123660 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168421 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149687 | BC117545.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149687 | BC118052.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149687 | AK077106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149687 | NM_177570.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149687 | BC117545.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149687 | BC118052.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149687 | AK077106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149687 | NM_177570.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23251744 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168421 | - | - |
| Whole Mouse Genome G4122A | A_52_P149687 | BC117545.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149687 | BC118052.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149687 | AK077106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149687 | NM_177570.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149687 | BC117545.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149687 | BC118052.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149687 | AK077106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149687 | NM_177570.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128571 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168421 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03280886 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03280894 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03280896 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03280867 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03280887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23251792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03614325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03614327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03280885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03280879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03614349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03280881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23251777 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10507723 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507721 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507725 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455838_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10507719 | - | - |
| Mouse Genome 430 2.0 Array | 1455838_at | - | - |
| Mouse Genome 430 2.0 Array | 1455838_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 194219_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 194219_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400010285.mm.1 | NM_177570.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010285.mm.1 | NM_177570.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010285.mm.1 | TC0400001352.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400010284.mm.1 | NM_177570.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010284.mm.1 | NM_177570.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010284.mm.1 | TC0400001352.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455838_at | - | - |
| Mouse Expression Set 430 | 1455838_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455838_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455838_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 404460 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2320750 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2320750 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002320750 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245376 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245376 | - | - |
| mouse-6 v1.1 (Illumina) | scl25014.5.1_128-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2320750 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25014.5.1_128-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320750 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1245376 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245376 | - | - |

Slfnl1
Genomic Map