Assay Details
| Gene Symbol: Gm12830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033617.1 | 1 - 2 | 193 | 59 | ||
| GenBank mRNA | AK038383.1 | - | 1 - 2 | 194 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 12829 | Gm12829 | OTTMUSG00000008582 | -138.3kb | Sense | Chr. 4 - 114680337 - 114683404 on NCBI Build GRCm38 |
| Downstream | forkhead box D2 | Foxd2 |
AI426778, Mf2 |
64.5kb | Anti-sense | Chr. 4 - 114906280 - 114908898 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67741 | NR_033617.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67741 | AK038383.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67741 | NR_033617.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P67741 | AK038383.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124354 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163800 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P67741 | NR_033617.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P67741 | AK038383.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P67741 | NR_033617.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P67741 | AK038383.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03947345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03647220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03683667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04006885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03263583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03409170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04006880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03647224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03409169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03710953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03290761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03207867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03647223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03273797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03273791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03273783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03409174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03290762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03710951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23221838 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.1 ST Array | 10507105 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400008794.mm.1 | NR_033617.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008794.mm.1 | NR_033617.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008794.mm.1 | TC0400001212.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400008792.mm.1 | NR_033617.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008792.mm.1 | NR_033617.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008792.mm.1 | TC0400001212.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400008793.mm.1 | NR_033617.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008793.mm.1 | NR_033617.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008793.mm.1 | TC0400001212.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400008795.mm.1 | NR_033617.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008795.mm.1 | NR_033617.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400008795.mm.1 | TC0400001212.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 313876 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 100580333 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234175 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100580333 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100580333 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234175 | - | - |

Gm12830
Genomic Map