Assay Details
| Gene Symbol: Gm31747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029727.1 | - | 2 - 3 | 739 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700065L07 gene | 1700065L07Rik | -118.4kb | Sense | Chr. 6 - 73436965 - 73471021 on NCBI Build GRCm38 | |
| Downstream |
catenin (cadherin associated protein), alpha 2 |
Ctnna2 |
AI481747, Catna, Catna2, cdf, chp |
3.3mb | Sense | Chr. 6 - 76881637 - 77979694 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31747
Genomic Map