Assay Details
| Gene Symbol: Gm20125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038020.1 | 1 - 2 | 334 | 84 | ||
| GenBank mRNA | AK029503.1 | - | 1 - 2 | 336 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA C330004P14 gene | C330004P14Rik | -1.2mb | Anti-sense | Chr. 10 - 15791777 - 15807065 on NCBI Build GRCm38 | |
| Downstream |
Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2 |
Cited2 |
AI835299, ER154-like, Mrg1, Msg2, p35srj |
627.4kb | Anti-sense | Chr. 10 - 17723205 - 17725674 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm20125
Genomic Map