Assay Details
| Gene Symbol: Nipal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK053937.1 | - | 1357 | 67 | ||
| AK082376.1 | - | 1355 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nipal3| Gene Symbol: Nipal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK053937.1 | - | 1357 | 67 | ||
| AK082376.1 | - | 1355 | 67 | |||
Set Membership: |
|