Assay Details
| Gene Symbol: Nipal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028995.3 | NP_083271.1 | 1 - 2 | 77 | 64 | |
| GenBank mRNA | AK018640.1 | - | 1 - 2 | 54 | 64 | |
| AK049696.1 | - | 1 - 2 | 112 | 64 | ||
| AK053937.1 | - | 80 | 64 | |||
| AK082376.1 | - | 79 | 64 | |||
| AK087811.1 | - | 1 - 2 | 80 | 64 | ||
| AK138843.1 | - | 1 - 2 | 78 | 64 | ||
| AK148206.1 | - | 1 - 2 | 96 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK018640.1, AK049696.1, AK082376.1, AK087811.1, AK138843.1, NM_028995.3 transcripts.
Set Membership: |
|

Nipal3
Genomic Map