Assay Details
| Gene Symbol: Nipal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028995.3 | NP_083271.1 | 2 - 3 | 426 | 150 | |
| GenBank mRNA | AK018640.1 | - | 2 - 3 | 403 | 150 | |
| AK049696.1 | - | 2 - 3 | 461 | 150 | ||
| AK053937.1 | - | 429 | 150 | |||
| AK082376.1 | - | 428 | 150 | |||
| AK087811.1 | - | 2 - 3 | 429 | 150 | ||
| AK138843.1 | - | 2 - 3 | 427 | 150 | ||
| AK148206.1 | - | 2 - 3 | 445 | 150 | ||
| BC057168.1 | - | 2 - 3 | 366 | 4973200 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nipal3
Genomic Map