Assay Details
| Gene Symbol: Nipal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028995.3 | NP_083271.1 | 8 - 9 | 1109 | 75 | |
| GenBank mRNA | AK018640.1 | - | 8 - 9 | 1086 | 75 | |
| AK049696.1 | - | 8 - 9 | 1144 | 75 | ||
| AK053937.1 | - | 1113 | 75 | |||
| AK087811.1 | - | 8 - 9 | 1112 | 75 | ||
| AK138843.1 | - | 8 - 9 | 1110 | 75 | ||
| AK148206.1 | - | 8 - 9 | 1130 | 75 | ||
| BC057168.1 | - | 8 - 9 | 1049 | 4973200 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nipal3
Genomic Map