Assay Details
| Gene Symbol: C87436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243741.1 | NP_001230670.1 | 3 - 4 | 1700 | 73 | |
| GenBank mRNA | AK077503.1 | - | 3 - 4 | 1587 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytotoxic granule-associated RNA binding protein 1 | Tia1 |
2310050N03Rik, AI256674, TIA-1, mTIA-1 |
-12.0kb | Sense | Chr. 6 - 86404219 - 86433405 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA A430078I02 gene | A430078I02Rik | T25621 | 23.4kb | Anti-sense | Chr. 6 - 86470786 - 86482352 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

C87436
Genomic Map