Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
TBC1 domain family, member 32 |
Tbc1d32 |
Bromi, D630037F22Rik, b2b2284Clo |
-148.0kb | Anti-sense | Chr. 10 - 56014293 - 56229456 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 36595 |
Gm36595 | 33.1kb | Sense | Chr. 10 - 56431504 - 56475419 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map