Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
gap junction protein, alpha 1 |
Gja1 |
AU042049, AW546267, Cnx43, Cx43, Cx43alpha1, Gja-1, Npm1, connexin43 |
-7.8kb | Sense | Chr. 10 - 56377201 - 56390423 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 36595 |
Gm36595 | 32.2kb | Sense | Chr. 10 - 56431504 - 56475419 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map