Assay Details
| Gene Symbol: Ankrd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023816.2 | NP_076305.2 | 4 - 5 | 847 | 90 | |
| GenBank mRNA | AF294328.1 | - | 4 - 5 | 849 | 90 | |
| AK005925.1 | - | 4 - 5 | 821 | 90 | ||
| AK132751.1 | - | 4 - 5 | 848 | 90 | ||
| AK134466.1 | - | 1 - 2 | 257 | 90 | ||
| BC120815.1 | - | 4 - 5 | 625 | 90 | ||
| BC120817.1 | - | 4 - 5 | 622 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd36
Genomic Map