Assay Details
| Gene Symbol: Ankrd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023816.2 | NP_076305.2 | 18 - 19 | 3003 | 64 | |
| GenBank mRNA | AK005775.1 | - | 3 - 4 | 811 | 64 | |
| AK132751.1 | - | 18 - 19 | 3002 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd36
Genomic Map