Assay Details
| Gene Symbol: Ankrd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023816.2 | NP_076305.2 | 19 - 20 | 3357 | 80 | |
| GenBank mRNA | AK005775.1 | - | 4 - 5 | 1165 | 80 | |
| AK132751.1 | - | 19 - 20 | 3356 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd36
Genomic Map