Assay Details
| Gene Symbol: Morc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159288.1 | NP_001152760.1 | 9 - 10 | 1426 | 104 | |
| NM_198162.1 | NP_937805.1 | 8 - 9 | 753 | 104 | ||
| GenBank mRNA | AK173042.1 | - | 9 - 10 | 1426 | 104 | |
| BC059243.1 | - | 8 - 9 | 753 | 6417551 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Morc2a
Genomic Map