Assay Details
| Gene Symbol: Fam50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138607.3 | NP_613073.2 | 10 - 11 | 1017 | 71 | |
| GenBank mRNA | AK008402.1 | - | 10 - 11 | 1018 | 71 | |
| BC026128.1 | - | 8 - 9 | 1570 | 71 | ||
| BC125297.1 | - | 9 - 10 | 883 | 40129812 | 71 | |
| Y18505.1 | - | 10 - 11 | 982 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam50a
Genomic Map