Assay Details
| Gene Symbol: Fam50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138607.3 | NP_613073.2 | 5 - 6 | 697 | 69 | |
| GenBank mRNA | AK008402.1 | - | 5 - 6 | 698 | 69 | |
| BC026128.1 | - | 4 - 5 | 1070 | 69 | ||
| BC125297.1 | - | 4 - 5 | 563 | 40129812 | 69 | |
| Y18505.1 | - | 5 - 6 | 662 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam50a
Genomic Map