Assay Details
| Gene Symbol: Arid1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080819.1 | NP_001074288.1 | 10 - 11 | 2985 | 65 | |
| GenBank mRNA | AK142143.1 | - | 9 - 10 | 2167 | 65 | |
| BC082554.1 | - | 2217 | 30622048 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arid1a
Genomic Map