| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400009283.mm.1 |
NM_008389.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009283.mm.1 |
NM_008389.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009283.mm.1 |
TC0400001253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400009285.mm.1 |
NM_008389.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009285.mm.1 |
NM_008389.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009285.mm.1 |
TC0400001253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400009287.mm.1 |
NM_008389.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009287.mm.1 |
NM_008389.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009287.mm.1 |
TC0400001253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400009288.mm.1 |
NM_008389.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009288.mm.1 |
NM_008389.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009288.mm.1 |
TC0400001253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400009286.mm.1 |
NM_008389.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009286.mm.1 |
NM_008389.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009286.mm.1 |
TC0400001253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400009289.mm.1 |
NM_008389.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009289.mm.1 |
NM_008389.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009289.mm.1 |
TC0400001253.mm.1 |
MTA TC |