Assay Details
| Gene Symbol: Ipp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008389.3 | NP_032415.2 | 5 - 6 | 1199 | 150 | |
| GenBank mRNA | AF285178.1 | - | 5 - 6 | 1089 | 150 | |
| AK131718.1 | - | 5 - 6 | 1200 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ipp
Genomic Map