Assay Details
| Gene Symbol: Mob3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175308.4 | NP_780517.1 | 1 - 2 | 149 | 71 | |
| GenBank mRNA | AK084014.1 | - | 1 - 2 | 150 | 71 | |
| AK148476.1 | - | 1 - 2 | 150 | 71 | ||
| BC099740.1 | - | 1 - 2 | 135 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK084014.1,AK148476.1,NM_175308.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mob3c
Genomic Map