Assay Details
| Gene Symbol: Tab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138667.3 | NP_619608.1 | 3 - 4 | 2012 | 62 | |
| GenBank mRNA | AB093262.1 | - | 2 - 3 | 1579 | 62 | |
| AK147830.1 | - | 3 - 4 | 1823 | 62 | ||
| AY093701.1 | - | 2 - 3 | 1672 | 62 | ||
| BC004072.1 | - | 1 - 2 | 161 | 62 | ||
| BC004813.1 | - | 2 - 3 | 1679 | 3586132 | 62 | |
More Information
Set Membership: |
|

Tab2
Genomic Map