Assay Details
| Gene Symbol: Tab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138667.3 | NP_619608.1 | 4 - 5 | 2175 | 100 | |
| GenBank mRNA | AB093262.1 | - | 3 - 4 | 1742 | 100 | |
| AK217747.1 | - | 1 - 2 | 125 | 100 | ||
| AY093701.1 | - | 3 - 4 | 1835 | 100 | ||
| BC004072.1 | - | 2 - 3 | 324 | 100 | ||
| BC004813.1 | - | 3 - 4 | 1842 | 3586132 | 100 | |
More Information
Set Membership: |
|

Tab2
Genomic Map