Assay Details
| Gene Symbol: Katna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011835.2 | NP_035965.2 | 6 - 7 | 868 | 105 | |
| GenBank mRNA | AF153197.1 | - | 6 - 7 | 846 | 105 | |
| AK012319.1 | - | 6 - 7 | 867 | 105 | ||
| AK041900.1 | - | 6 - 7 | 863 | 105 | ||
| BC009136.1 | - | 6 - 7 | 782 | 3597742 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Katna1
Genomic Map