Assay Details
| Gene Symbol: Zc3h12d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172785.3 | NP_766373.2 | 3 - 4 | 619 | 113 | |
| GenBank mRNA | AK052806.1 | - | 3 - 4 | 621 | 113 | |
| BC112398.1 | - | 2 - 3 | 401 | 40062049 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zc3h12d
Genomic Map