Assay Details
| Gene Symbol: Lats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010690.1 | NP_034820.1 | 6 - 7 | 3143 | 99 | |
| GenBank mRNA | AF104414.1 | - | 3 - 4 | 2268 | 99 | |
| BC158092.1 | - | 5 - 6 | 2989 | 9088177 | 99 | |
| BC158123.1 | - | 6 - 7 | 3138 | 9088196 | 99 | |
| CO045800.1 | - | 2 - 3 | 206 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lats1
Genomic Map