Assay Details
| Gene Symbol: Npc1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207242.2 | NP_997125.2 | 16 - 17 | 3641 | 61 | |
| GenBank mRNA | AK078947.1 | - | 16 - 17 | 3542 | 61 | |
| AY437866.1 | - | 16 - 17 | 3519 | 61 | ||
| BC131788.1 | - | 16 - 17 | 3518 | 61 | ||
| BC131789.1 | - | 8 - 9 | 2483 | 40086103 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Npc1l1
Genomic Map