Assay Details
| Gene Symbol: E2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK087452.1 | - | 1 - 2 | 643 | 65 | |
| BC062101.1 | - | 1 - 2 | 350 | 6848237 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

E2f2
Genomic Map