Assay Details
| Gene Symbol: Ppie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019489.5 | NP_062362.1 | 7 - 8 | 521 | 86 | |
| GenBank mRNA | AF182825.1 | - | 7 - 8 | 503 | 86 | |
| AK008177.1 | - | 7 - 8 | 522 | 86 | ||
| AK042165.1 | - | 4 - 5 | 3105 | 86 | ||
| BC045154.1 | - | 7 - 8 | 516 | 6468589 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppie
Genomic Map