Assay Details
| Gene Symbol: Ermap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013848.2 | NP_038876.2 | 6 - 7 | 1122 | 71 | |
| GenBank mRNA | AF153906.1 | - | 6 - 7 | 1087 | 71 | |
| BC063257.1 | - | 6 - 7 | 1056 | 6410069 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ermap
Genomic Map