Assay Details
| Gene Symbol: Kcnmb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031169.4 | NP_112446.2 | 1 - 2 | 309 | 108 | |
| GenBank mRNA | AF020711.1 | - | 1 - 2 | 309 | 108 | |
| BC013338.1 | - | 1 - 2 | 100 | 3988005 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF020711.1, BC013338.1, NM_031169.4 transcripts.
Set Membership: |
|

Kcnmb1
Genomic Map