Assay Details
| Gene Symbol: Mpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010822.3 | NP_034952.2 | 1 - 2 | 214 | 73 | |
| GenBank mRNA | AK018694.1 | - | 1 - 2 | 194 | 73 | |
| AK076201.1 | - | 1 - 2 | 209 | 73 | ||
| AK088878.1 | - | 1 - 2 | 168 | 73 | ||
| AK134900.1 | - | 1 - 2 | 181 | 73 | ||
| AK155766.1 | - | 1 - 2 | 215 | 73 | ||
| AK156067.1 | - | 1 - 2 | 181 | 73 | ||
| BC014754.1 | - | 1 - 2 | 208 | 4486938 | 73 | |
| BY309585.1 | - | 1 - 2 | 214 | 73 | ||
| U10420.1 | - | 1 - 2 | 207 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpg
Genomic Map