Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100022975.mm.1 |
NM_001134458.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022975.mm.1 |
NM_001134458.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022975.mm.1 |
NM_008374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022975.mm.1 |
NM_008374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022975.mm.1 |
TC1100002470.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100022974.mm.1 |
NM_001134458.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022974.mm.1 |
NM_001134458.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022974.mm.1 |
NM_008374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022974.mm.1 |
NM_008374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022974.mm.1 |
TC1100002470.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100022976.mm.1 |
NM_001134458.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022976.mm.1 |
NM_001134458.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022976.mm.1 |
NM_008374.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022976.mm.1 |
NM_008374.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100022976.mm.1 |
TC1100002470.mm.1 |
MTA TC |