Assay Details
| Gene Symbol: Rhbdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291818.1 | NP_001278747.1 | 6 - 7 | 826 | 79 | |
| NM_010117.2 | NP_034247.2 | 6 - 7 | 945 | 79 | ||
| GenBank mRNA | BC023469.1 | - | 1 - 2 | 93 | 79 | |
| BC027346.1 | - | 6 - 7 | 890 | 4011421 | 79 | |
| CD554689.1 | - | 2 - 3 | 243 | 79 | ||
| CN664154.1 | - | 3 - 4 | 526 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rhbdf1
Genomic Map