Assay Details
| Gene Symbol: Rhbdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291818.1 | NP_001278747.1 | 8 - 9 | 1243 | 59 | |
| NM_010117.2 | NP_034247.2 | 8 - 9 | 1362 | 59 | ||
| GenBank mRNA | AK144437.1 | - | 8 - 9 | 1231 | 59 | |
| AK220566.1 | - | 5 - 6 | 2778 | 59 | ||
| BC023469.1 | - | 3 - 4 | 510 | 59 | ||
| BC027346.1 | - | 8 - 9 | 1307 | 4011421 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhbdf1
Genomic Map