Assay Details
| Gene Symbol: Abcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007435.1 | NP_031461.1 | 8 - 9 | 2313 | 63 | |
| GenBank mRNA | AK088792.1 | - | 8 - 9 | 2183 | 63 | |
| BC011273.1 | - | 8 - 9 | 2132 | 3710503 | 63 | |
| BC079840.1 | - | 8 - 9 | 2191 | 6810147 | 63 | |
| Z33637.1 | - | 8 - 9 | 2313 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcd1
Genomic Map