Assay Details
| Gene Symbol: Traf3ip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134000.3 | NP_598761.2 | 7 - 8 | 1866 | 96 | |
| GenBank mRNA | AB238206.1 | - | 6 - 7 | 1445 | 96 | |
| AB238207.1 | - | 6 - 7 | 1445 | 96 | ||
| AK031556.1 | - | 7 - 8 | 1591 | 96 | ||
| AK037407.1 | - | 7 - 8 | 1673 | 96 | ||
| AK052190.1 | - | 7 - 8 | 1866 | 96 | ||
| AK098107.1 | - | 7 - 8 | 1588 | 96 | ||
| BC008534.1 | - | 6 - 7 | 1022 | 96 | ||
| BC008535.1 | - | 6 - 7 | 1022 | 96 | ||
| BC022633.1 | - | 4 - 5 | 292 | 96 | ||
| BC071207.1 | - | 4 - 5 | 360 | 96 | ||
| BC096483.1 | - | 7 - 8 | 1547 | 5310588 | 96 | |
| BC138234.1 | - | 7 - 8 | 1636 | 8861253 | 96 | |
| BC138235.1 | - | 7 - 8 | 1636 | 8861254 | 96 | |
| JX869941.1 | - | 7 - 8 | 1873 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Traf3ip2
Genomic Map