Assay Details
| Gene Symbol: Gtf3c6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026113.4 | NP_080389.2 | 5 - 6 | 389 | 79 | |
| GenBank mRNA | AK005261.1 | - | 5 - 6 | 388 | 79 | |
| AK007816.1 | - | 5 - 6 | 390 | 79 | ||
| AK010514.1 | - | 5 - 6 | 389 | 79 | ||
| AK162053.1 | - | 5 - 6 | 389 | 79 | ||
| AK168070.1 | - | 5 - 6 | 388 | 79 | ||
| AK185766.1 | - | 1 - 2 | 98 | 79 | ||
| BC048446.1 | - | 5 - 6 | 395 | 6590885 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27083090 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27083091 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27083092 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963777 | AK162053.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963777 | AK168070.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963777 | BC027260.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963777 | NM_026113.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963777 | AK005261.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758679 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918372 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914614 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121694 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150380 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963777 | AK162053.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963777 | AK168070.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963777 | BC027260.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963777 | NM_026113.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963777 | AK005261.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27083065 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07035769 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150380 | - | - |
| Whole Mouse Genome G4122A | A_51_P382732 | AK010514.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627793 | AK010514.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627793 | AK162053.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627793 | AK168070.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627793 | BC048446.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627793 | NM_026113.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127305 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150380 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08945471 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08945470 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08525183 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08945456 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27083091 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27083092 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27083090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27082999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27083048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08945464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08735771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08765046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08654734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08765045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08654737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08378393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08735773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08938202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08654736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27083054 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10368741 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368744 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368743 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368746 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368747 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368745 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420626_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420625_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10368739 | - | - |
| Mouse Genome 430 2.0 Array | 1420625_at | - | - |
| Mouse Genome 430 2.0 Array | 1434568_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434568_at | - | - |
| Mouse Genome 430 2.0 Array | 1420626_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420625_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420626_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67371_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67371_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000014496.mm.1 | NM_026113.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014496.mm.1 | NM_026113.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014496.mm.1 | TC1000002042.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000014497.mm.1 | NM_026113.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014497.mm.1 | NM_026113.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014497.mm.1 | TC1000002042.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420626_PM_at | - | - |
| Mouse Expression Set 430 | 1420625_at | - | - |
| Mouse Expression Set 430 | 1420625_PM_at | - | - |
| Mouse Expression Set 430 | 1420626_at | - | - |
| Mouse Expression Set 430 | 1434568_at | - | - |
| Mouse Expression Set 430 | 1434568_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420625_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420626_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420625_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420626_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420626_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420626_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420625_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420625_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 436091 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2100215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 360546 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002100215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000360546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245249 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257110 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245249 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257110 | - | - |
| mouse-6 v1.1 (Illumina) | scl38030.6.1_17-S | - | - |
| mouse-6 v1.1 (Illumina) | scl067371.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 360546 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2100215 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067371.1_2-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38030.6.1_17-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100215 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 360546 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2940399 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2940394 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1257110 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257110 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2940394 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2940399 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl38030.6.1_17-S | - | - |

Gtf3c6
Genomic Map