Assay Details
| Gene Symbol: Enpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134005.2 | NP_598766.2 | 8 - 9 | 863 | 127 | |
| GenBank mRNA | AK132256.1 | - | 8 - 9 | 876 | 127 | |
| BC117533.1 | - | 8 - 9 | 781 | 127 | ||
| BC118512.1 | - | 8 - 9 | 781 | 127 | ||
More Information
Set Membership: |
|

Enpp3
Genomic Map