Assay Details
| Gene Symbol: Enpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF339910.1 | - | 9 - 10 | 969 | 89 | |
| AK029600.1 | - | 9 - 10 | 881 | 89 | ||
| AK036952.1 | - | 9 - 10 | 978 | 89 | ||
| AK041894.1 | - | 4 - 5 | 342 | 89 | ||
| AK088857.1 | - | 9 - 10 | 976 | 89 | ||
| AK145233.1 | - | 9 - 10 | 972 | 89 | ||
| AK164619.1 | - | 9 - 10 | 977 | 89 | ||
| BC050011.1 | - | 9 - 10 | 1149 | 89 | ||
| J02700.2 | - | 9 - 10 | 1080 | 89 | ||
More Information
Set Membership: |
|

Enpp1
Genomic Map