Assay Details
| Gene Symbol: Slc6a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008135.4 | NP_032161.2 | 8 - 9 | 1267 | 75 | |
| GenBank mRNA | AK014572.1 | - | 1111 | 75 | ||
| AK048168.1 | - | 1152 | 75 | |||
| BC021828.1 | - | 8 - 9 | 1090 | 4194951 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a9
Genomic Map