Assay Details
| Gene Symbol: Dph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026344.3 | NP_080620.1 | 2 - 3 | 409 | 108 | |
| GenBank mRNA | AK011199.1 | - | 2 - 3 | 427 | 108 | |
| AK018638.1 | - | 2 - 3 | 410 | 108 | ||
| AK051026.1 | - | 2 - 3 | 428 | 108 | ||
| AK080747.1 | - | 2 - 3 | 427 | 108 | ||
| BC031124.1 | - | 2 - 3 | 396 | 5029876 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dph2
Genomic Map