Assay Details
| Gene Symbol: Dph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026344.3 | NP_080620.1 | 5 - 6 | 1492 | 74 | |
| GenBank mRNA | AK011199.1 | - | 5 - 6 | 1510 | 74 | |
| AK018638.1 | - | 5 - 6 | 1493 | 74 | ||
| AK051026.1 | - | 5 - 6 | 1512 | 74 | ||
| AK080747.1 | - | 3 - 4 | 1789 | 74 | ||
| BC014746.1 | - | 4 - 5 | 1388 | 74 | ||
| BC031124.1 | - | 5 - 6 | 1479 | 5029876 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23237104 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23237102 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23237103 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440892 | BC031124.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440892 | AK080747.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440892 | AK194858.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440892 | AK218933.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440892 | AK018638.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440892 | BC031124.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440892 | AK080747.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440892 | AK194858.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440892 | AK218933.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440892 | AK018638.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526948 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955937 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142163 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440892 | BC031124.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440892 | AK080747.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440892 | AK194858.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440892 | AK218933.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440892 | AK018638.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440892 | BC031124.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440892 | AK080747.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440892 | AK194858.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440892 | AK218933.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440892 | AK018638.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440892 | BC031124.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440892 | AK080747.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440892 | AK194858.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440892 | AK218933.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440892 | AK018638.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440892 | BC031124.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440892 | AK080747.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440892 | AK194858.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440892 | AK218933.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440892 | AK018638.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23237091 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142163 | - | - |
| Whole Mouse Genome G4122A | A_51_P440892 | BC031124.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440892 | AK080747.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440892 | AK194858.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440892 | AK218933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440892 | AK018638.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440892 | BC031124.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440892 | AK080747.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440892 | AK194858.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440892 | AK218933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440892 | AK018638.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101602 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142163 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03677622 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03315718 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03315716 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03315714 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03821600 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03821601 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23237102 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23237104 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23237103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23237094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03315716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23237081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03489335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23237120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23237105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23237093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23237085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23237115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23237118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03950128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23237083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03821600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03677622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03821601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03315714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03489334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03315717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23237114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03489338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03902601 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10515530 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515535 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515529 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515533 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515531 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515532 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515534 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452647_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10515528 | - | - |
| Mouse Genome 430 2.0 Array | 1452647_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1452647_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67728_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67728_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400027193.mm.1 | NM_026344.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027193.mm.1 | NM_026344.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027193.mm.1 | TC0400003380.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400027190.mm.1 | NM_026344.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027190.mm.1 | NM_026344.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027190.mm.1 | TC0400003380.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452647_a_at | - | - |
| Mouse Expression Set 430 | 1452647_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452647_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452647_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452647_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452647_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2100402 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3060039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3060039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2100402 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002100402 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003060039 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2701741 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2613389 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2701741 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2613389 | - | - |
| mouse-6 v1.1 (Illumina) | scl23928.4.1_17-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0002649.1_140-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2100402 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3060039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002649.1_140-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23928.4.1_17-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3060039 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100402 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2847024 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2613389 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2701741 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2847024 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2613389 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2701741 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl23928.4.1_17-S | - | - |

Dph2
Genomic Map